Polyethylene glycol mono-4-octylphenyl ether, M.W. n=∼10 for biochemical research

Supplier: TCI America

P1775-25G P1775-5G
TCP1775-25GEA 72.68 CAD
TCP1775-25G TCP1775-5G
Polyethylene glycol mono-4-octylphenyl ether, M.W. n=∼10 for biochemical research
Polyethylene glycol mono-4-octylphenyl ether

[for Biochemical Research]
CAS Number: 9002-93-1
MDL Number: MFCD00132505
Form: Clear Liquid
Color: Colorless
Melting point (°C): 6
Flash Point (°C): 286
Specific Gravity (20/20): 1.07


Product Categories: Bioscience, Nonionic Surfactants (for Biochemistry)

Formula: C8H17C6H4(OCH2CH2)nOH
Melting Pt: 6 °C
Density: 1.07 (20 °C)
Flash Pt: 286 °C
MDL Number: MFCD00132505
CAS Number: 9002-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e605b52cf895457becb041f14a17296b1bded0ca&vid=c11f1c579b72805c386a1aae62dfb3a002686859&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR