L-Proline-tert-butyl ester hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
P1729-25G
P1729-5G
TCP1729-25GEA
451.88
CAD
TCP1729-25G
TCP1729-5G
L-Proline-tert-butyl ester hydrochloride ≥98.0% (by total nitrogen and titration analysis)
L-Proline-tert-butyl ester hydrochloride
CAS Number: 5497-76-7
MDL Number: MFCD00153459
Molecular Formula: C9H17NO2
Molecular Weight: 207.70
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Color: White
Melting point (°C): 112
Specific rotation [a]20/D: -32 deg (C=1, H2O)
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Pro-OtBu.HCl
Formula:
C₉H₁₇NO₂ Melting Pt: 112 °C |
MDL Number:
MFCD00153459 CAS Number: 5497-76-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...