Phenyl-3,4,6-tri-O-acetyl-2-deoxy-1-thio-2-(2,2,2-trichloroethoxyformamido)-β-D-galactopyranoside ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
P1642-1G
P1642-5G
TCP1642-1GEA
373.51
CAD
TCP1642-1G
TCP1642-5G
Phenyl-3,4,6-tri-O-acetyl-2-deoxy-1-thio-2-(2,2,2-trichloroethoxyformamido)-β-D-galactopyranoside ≥98.0% (by HPLC, total nitrogen)
Phenyl-3,4,6-tri-O-acetyl-2-deoxy-1-thio-2-(2,2,2-trichloroethoxyformamido)-beta-D-galactopyranoside
CAS Number: 278784-83-1
Molecular Formula: C21H24Cl3NO9S
Molecular Weight: 572.83
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 117
Specific rotation [a]20/D: -5 deg (C=1, CHCl3)
Storage Temperature: 0-10°C
Formula:
C₂₁H₂₄Cl₃NO₉S MW: 572.83 g/mol Melting Pt: 117 °C |
CAS Number:
278784-83-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...