Potassium-tert-butoxide 12% ∼1 mol/L in THF

Supplier: TCI America

P1619-100ML
TCP1619-100MLEA 99.73 CAD
TCP1619-100ML
Potassium-tert-butoxide 12% ∼1 mol/L in THF
Potassium-tert-butoxide

CAS Number: 865-47-4
Molecular Formula: C4H9KO
Molecular Weight: 112.21
Form: Clear Liquid
Color: Very Pale Yellow
Flash Point (°C): -19
Specific Gravity (20/20): 0.91

Formula: C₄H₉KO
Density: 0.91 (20 °C)
Flash Pt: –19 °C
MDL Number: MFCD00012162
CAS Number: 865-47-4
UN: 3206
ADR: 4.2,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbcb37d9a905b31ef1f1e99a5cef48ef4f7af662&vid=9e1a173e0bdce6dd3cd4aaae7a899410228ee46b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR