Palladium 10% on carbon wetted with ca. 55% water
Supplier: TCI America
P1491-25G
P1491-5G
TCP1491-25GEA
826.81
CAD
TCP1491-25G
TCP1491-5G
Palladium 10% on carbon wetted with ca. 55% water
Palladium
CAS Number: 7440-05-3
Molecular Formula: Pd
Molecular Weight: 106.42
Form: Powder
Color: Black
Product Categories: Synthetic Organic Chemistry, Functional Group Transformation and Introducing Reaction, Metal Catalysts for Organic Synthesis, Suzuki-Miyaura Cross-coupling Reaction
Formula:
Pd |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...