Palladium 5% on carbon wetted with ca. 55% water

Supplier: TCI America

P1490-25G P1490-5G
TCP1490-25GEA 434.5 CAD
TCP1490-25G TCP1490-5G
Palladium 5% on carbon wetted with ca. 55% water
Palladium

CAS Number: 7440-05-3
Molecular Formula: Pd
Molecular Weight: 106.42
Form: Crystals
Color: Black


Product Categories: Synthetic Organic Chemistry, Functional Group Transformation and Introducing Reaction, Metal Catalysts for Organic Synthesis, Suzuki-Miyaura Cross-coupling Reaction

Formula: Pd
MDL Number: MFCD00011167
CAS Number: 7440-05-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cf2d7065894664d416b06f556ea7b64428b28f&vid=1e465d6587e800ac5554b1a1097150ba6915a7eb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR