Palladium(II) chloride ≥98.0%

Supplier: TCI America

Synonyms: Palladium dichloride

P1489-1G P1489-5G
TCP1489-1GEA 525.67 CAD
TCP1489-1G TCP1489-5G
Palladium(II) chloride ≥98.0%
Palladium (II) chloride

CAS Number: 7647-10-1
MDL Number: MFCD00003558
Molecular Formula: Cl2Pd
Molecular Weight: 177.32
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Deep Yellow Red
Melting point (°C): 680


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: Palladium Dichloride

Formula: PdCl₂
MW: 177.33 g/mol
Melting Pt: 678
MDL Number: MFCD00003558
CAS Number: 7647-10-1
UN: 3260
ADR: 8 III
Merck Index: 13,07058
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bb455168f0c9940085f225e032b59f2cc5edf03&vid=2bb455168f0c9940085f225e032b59f2cc5edf03&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR