4,4,5,5,5-Pentafluoropentan-1-ol ≥93.0% (by GC)

Supplier: TCI America

P1342-25G P1342-5G
TCP1342-25GEA 285.98 CAD
TCP1342-25G TCP1342-5G
4,4,5,5,5-Pentafluoropentan-1-ol ≥93.0% (by GC)
4,4,5,5,5-Pentafluoropentan-1-ol

CAS Number: 148043-73-6
MDL Number: MFCD00153224
Molecular Formula: C5H7F5O
Molecular Weight: 178.10
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 135
Flash Point (°C): 63
Specific Gravity (20/20): 1.35

Formula: C₅H₇F₅O
Boiling Pt: 135 °C
Density: 1.35 (20 °C)
Flash Pt: 63 °C
MDL Number: MFCD00153224
CAS Number: 148043-73-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acb54b31d93c662fabdd10e086ba84d833bd0431&vid=acb54b31d93c662fabdd10e086ba84d833bd0431&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR