Pentafluorophenol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,3,4,5,6-Pentafluorophenol

P0919-10G P0919-25G
TCP0919-10GEA 82.63 CAD
TCP0919-10G TCP0919-25G
Pentafluorophenol ≥98.0% (by GC)
Pentafluorophenol

CAS Number: 771-61-9
MDL Number: MFCD00002156
Molecular Formula: C6HF5O
Molecular Weight: 184.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 143
Flash Point (°C): 72
Freezing point (°C): 36


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Peptide Synthesis
Other Product Names / Synonyms: Perfluorophenol

Formula: C₆HF₅O
MW: 184.07
Boiling Pt: 143 °C
Density: 1.75
Flash Pt: 72 °C
MDL Number: MFCD00002156
CAS Number: 771-61-9
UN: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8c19b71ae36c0ee3c8e28f6e0ff33015ac49c42&vid=d326089f32e60be7faafca430c5ef7a9f9bf1914&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR