Polyethylene glycol monooleyl ether, M.W. n=∼2
Supplier: TCI America
P0711-25ML
P0711-500ML
TCP0711-25MLEA
30.02
CAD
TCP0711-25ML
TCP0711-500ML
Polyethylene glycol monooleyl ether, M.W. n=∼2
Polyethylene glycol monooleyl ether
CAS Number: 9004-98-2
MDL Number: MFCD00074369
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 110
Specific Gravity (20/20): 0.90
Product Categories: Materials Chemistry, Nonionic Surfactants
Formula:
HO(CH2CH2O)nC18H35 Density: 0.90 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00074369 CAS Number: 9004-98-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...