Tris(perfluorobutyl)amine 60.0% (by GC)
Supplier: TCI America
Synonyms:
Perfluorotri-n-butylamine, Heptacosafluorotri-n-butylamine, Perfluorotributylamine, FC-43, Heptacosafluorotributylamine, PFTBA, Tris(nonafluorobutyl)amine
[for Mass spectrometry]
CAS Number: 311-89-7
MDL Number: MFCD00000436
Molecular Formula: C12F27N
Molecular Weight: 671.10
Purity/Analysis Method: >60.0% (GC)
Form: Clear Liquid
Boiling point (°C): 178
Specific Gravity (20/20): 1.89
Product Categories: Synthetic Organic Chemistry, Fluorous Chemistry, Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: Perfluorotributylamine, Tris(nonafluorobutyl)amine
Formula:
C₁₂F₂₇N MW: 671.1 g/mol Boiling Pt: 178 °C Density: 1.89 (20 °C) |
MDL Number:
MFCD00000436 CAS Number: 311-89-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...