3-Oxo-N-(4-trifluoromethylphenyl)butyramide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 4'-(Trifluoromethyl)acetoacetanilide, 3-Oxo-N-(4-(trifluoromethyl)phenyl)butanamide

O0444-1G O0444-200MG
TCO0444-1GEA 636.42 CAD
TCO0444-1G TCO0444-200MG
3-Oxo-N-(4-trifluoromethylphenyl)butyramide ≥98.0% (by HPLC, total nitrogen)
3-Oxo-N-(4-trifluoromethylphenyl)butyramide

CAS Number: 351-87-1
MDL Number: MFCD00231301
Molecular Formula: C11H10F3NO2
Molecular Weight: 245.20
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 154


Product Categories: Synthetic Organic Chemistry, Diketone Compounds (Multi-component Reaction), ƒÀ-Keto Esters (Multi-component Reaction), Fluorinated Building Blocks
Other Product Names / Synonyms: 4'-(Trifluoromethyl)acetoacetanilide

Formula: C₁₁H₁₀F₃NO₂
MW: 245.20 g/mol
Melting Pt: 154 °C
MDL Number: MFCD00231301
CAS Number: 351-87-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79d4b6dc11734346fce3bdfff23cc14007ad36b7&vid=79d4b6dc11734346fce3bdfff23cc14007ad36b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR