1-[2-Oxo-2-(2-pyridyl)ethyl]pyridinium iodide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

O0438-1G O0438-5G
TCO0438-1GEA 42.66 CAD
TCO0438-1G TCO0438-5G
1-[2-Oxo-2-(2-pyridyl)ethyl]pyridinium iodide ≥98.0% (by HPLC, titration analysis)
1-[2-Oxo-2-(2-pyridyl)ethyl]pyridinium iodide

CAS Number: 26482-00-8
MDL Number: MFCD01690884
Molecular Formula: C12H11IN2O
Molecular Weight: 326.14
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 190
Storage Temperature: 0-10°C

Formula: C₁₂H₁₁IN₂O
Melting Pt: 190 °C
MDL Number: MFCD01690884
CAS Number: 26482-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6b40c7dbfbda21a624c6dfb7f7bd2607058dbbd&vid=d6b40c7dbfbda21a624c6dfb7f7bd2607058dbbd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR