3-Oxabicyclo[3.1.0]hexane-2,4-dione ≥98.0%

Supplier: TCI America

Synonyms: Cyclopropane-1,2-dicarboxylic acid anhydride

O0434-1G O0434-5G
TCO0434-1GEA 45.82 CAD
TCO0434-1G TCO0434-5G
3-Oxabicyclo[3.1.0]hexane-2,4-dione ≥98.0%
3-Oxabicyclo[3.1.0]hexane-2,4-dione

CAS Number: 5617-74-3
MDL Number: MFCD00126929
Molecular Formula: C5H4O3
Molecular Weight: 112.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 114
Melting point (°C): 60


Other Product Names / Synonyms: 1,2-Cyclopropanedicarboxylic Anhydride

Formula: C5H4O3
Boiling Pt: 114 °C
Melting Pt: 60 °C
MDL Number: MFCD00126929
CAS Number: 5617-74-3

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR