9-Oxoacridine-10-acetic acid (CMA) ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

O0432-1G O0432-5G
TCO0432-1GEA 115.66 CAD
TCO0432-1G TCO0432-5G
9-Oxoacridine-10-acetic acid (CMA) ≥98.0% (by HPLC, titration analysis)
9-Oxoacridine-10-acetic acid (CMA)

CAS Number: 38609-97-1
MDL Number: MFCD00157050
Molecular Formula: C15H11NO3
Molecular Weight: 253.26
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 291
Lambda max.: 410 nm (H2O)


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 10-(Carboxymethyl)-9(10H)-acridone, 9-Oxo-10(9H)-acridineacetic Acid, Cridanimod

Formula: C₁₅H₁₁NO₃
Melting Pt: 291 °C
MDL Number: MFCD00157050
CAS Number: 38609-97-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6423779eb029b519c999c33f034baca01dd91f16&vid=6423779eb029b519c999c33f034baca01dd91f16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR