4-n-Octyl-4H-dithieno[3,2-b:2',3'-d]pyrrole ≥98.0% (by GC)
Supplier: TCI America
O0431-1G
O0431-200MG
TCO0431-1GEA
699.15
CAD
TCO0431-1G
TCO0431-200MG
4-n-Octyl-4H-dithieno[3,2-b:2',3'-d]pyrrole ≥98.0% (by GC)
4-n-Octyl-4H-dithieno[3,2-b:2',3'-d]pyrrole
CAS Number: 141029-75-6
MDL Number: MFCD26793506
Molecular Formula: C16H21NS2
Molecular Weight: 291.47
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 35
Storage Temperature: 0-10°C
Formula:
C₁₆H₂₁NS₂ Melting Pt: 35 °C |
MDL Number:
MFCD26793506 CAS Number: 141029-75-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...