9-(9-Heptadecanyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥97.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
9-(9-Heptadecanyl)carbazole-2,7-diboronic Acid Bis(pinacol) Ester, 9-(1-n-Octylnonyl)carbazole-2,7-diboronic Acid Bis(pinacol) Ester, 9-(Heptadecan-9-yl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole, 9-(1-n-Octylnonyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
CAS Number: 958261-51-3
MDL Number: MFCD16621134
Molecular Formula: C41H65B2NO4
Molecular Weight: 657.59
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Crystal
Melting point (°C): 146
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives, Materials Chemistry, Building Blocks for Semiconducting Polymers, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 9-(1-n-Octylnonyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole, 9-(1-n-Octylnonyl)carbazole-2,7-diboronic Acid Bis(pinacol) Ester, 9-(9-Heptadecanyl)carbazole-2,7-diboronic Acid Bis(pinacol) Ester
Formula:
C₄₁H₆₅B₂NO₄ MW: 657.59 g/mol Melting Pt: 146 °C |
MDL Number:
MFCD16621134 CAS Number: 958261-51-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...