Ozagrel hydrochloride hydrate ≥98.0%
Supplier: TCI America
Synonyms:
Ozagrel HCl, (E)-3-(4-((1H-Imidazol-1-yl)methyl)phenyl)acrylic acid hydrochloride, trans-4-[(1-Imidazolyl)methyl]cinnamic Acid Hydrochloride, (E)-3-[4-(Imidazol-1-ylmethyl)phenyl]propenoic Acid Hydrochloride
CAS Number: 78712-43-3
MDL Number: MFCD06795644
Molecular Formula: C13H12N2O2
Molecular Weight: 264.71
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 217
Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Antithrombotic Ingredients
Other Product Names / Synonyms: (E)-3-[4-(Imidazol-1-ylmethyl)phenyl]propenoic Acid Hydrochloride, trans-4-[(1-Imidazolyl)methyl]cinnamic Acid Hydrochloride
Formula:
C₁₃H₁₂N₂O₂ MW: 264.71 g/mol Melting Pt: 217 °C |
MDL Number:
MFCD06795644 CAS Number: 78712-43-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...