3-n-Octyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4,4,5,5-Tetramethyl-2-(3-n-octyl-2-thienyl)-1,3,2-dioxaborolan, 3-n-Octyl-2-thiopheneboronic Acid Pinacol Ester
CAS Number: 405165-14-2
Molecular Formula: C18H31BO2S
Molecular Weight: 322.31
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.98
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives, Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 4,4,5,5-Tetramethyl-2-(3-n-octyl-2-thienyl)-1,3,2-dioxaborolan, 3-n-Octyl-2-thiopheneboronic Acid Pinacol Ester
Formula:
C₁₈H₃₁BO₂S MW: 322.31 g/mol Density: 0.98 (20 °C) |
CAS Number:
405165-14-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...