Oxetan-3-ol ≥96.0%

Supplier: TCI America

Synonyms: 3-Hydroxyoxetane

O0409-1G O0409-5G
TCO0409-1GEA 44.24 CAD
TCO0409-1G TCO0409-5G
Oxetan-3-ol ≥96.0%
Oxetan-3-ol

CAS Number: 7748-36-9
MDL Number: MFCD09056790
Molecular Formula: C3H6O2
Molecular Weight: 74.08
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 50
Flash Point (°C): 87
Specific Gravity (20/20): 1.14
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3-Hydroxyoxetane, 1,3-Epoxy-2-propanol

Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 50 °C
Density: 1.14 (20 °C)
Flash Pt: 87 °C
MDL Number: MFCD09056790
CAS Number: 7748-36-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c84d00378fdd04085b46088403655439c631901&vid=9c84d00378fdd04085b46088403655439c631901&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR