Oxiracetam ≥96.0% (by GC, total nitrogen)

Supplier: TCI America

O0398-1G O0398-5G
TCO0398-1GEA 142.2 CAD
TCO0398-1G TCO0398-5G
Oxiracetam ≥96.0% (by GC, total nitrogen)
Oxiracetam

CAS Number: 62613-82-5
MDL Number: MFCD00242951
Molecular Formula: C6H10N2O3
Molecular Weight: 158.16
Purity/Analysis Method: >96.0% (GC,N)
Form: Crystal
Melting point (°C): 172
Storage Temperature: 0-10°C


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 2-(4-Hydroxy-2-oxo-1-pyrrolidinyl)acetamide

Formula: C₆H₁₀N₂O₃
Melting Pt: 172 °C
MDL Number: MFCD00242951
CAS Number: 62613-82-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=323e52482097e54f2c0ddb54edf3133bde22dabc&vid=323e52482097e54f2c0ddb54edf3133bde22dabc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR