1,1,1,3,5,7,7,7-Octamethyltetrasiloxane ≥95.0% (by GC)

Supplier: TCI America

O0397-25G O0397-5G
TCO0397-25GEA 202.52 CAD
TCO0397-25G TCO0397-5G
1,1,1,3,5,7,7,7-Octamethyltetrasiloxane ≥95.0% (by GC)
1,1,1,3,5,7,7,7-Octamethyltetrasiloxane

CAS Number: 16066-09-4
MDL Number: MFCD00053664
Molecular Formula: C8H26O3Si4
Molecular Weight: 282.63
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 177
Flash Point (°C): 63
Specific Gravity (20/20): 0.86

Formula: C₈H₂₆O₃Si₄
Boiling Pt: 177 °C
Density: 0.86 (20 °C)
Flash Pt: 63 °C
MDL Number: MFCD00053664
CAS Number: 16066-09-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f145bbe1dd63d1e75c1876574cc6339fd8ff7125&vid=f145bbe1dd63d1e75c1876574cc6339fd8ff7125&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR