N-n-Octyl-3,4-thiophenedicarboximide ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
5-n-Octyl-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione, 5-Octyl-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione
CAS Number: 773881-43-9
Molecular Formula: C14H19NO2S
Molecular Weight: 265.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 124
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 5-n-Octyl-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione
Formula:
C₁₄H₁₉NO₂S MW: 265.37 g/mol Melting Pt: 124 °C |
MDL Number:
MFCD23703118 CAS Number: 773881-43-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...