3-Oxocyclobutanecarboxylic acid ≥97.0%
Supplier: TCI America
O0392-1G
O0392-5G
TCO0392-1GEA
33.18
CAD
TCO0392-1G
TCO0392-5G
3-Oxocyclobutanecarboxylic acid ≥97.0%
3-Oxocyclobutanecarboxylic acid
CAS Number: 23761-23-1
MDL Number: MFCD00100900
Molecular Formula: C5H6O3
Molecular Weight: 114.10
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 70
Formula:
C5H6O3 Melting Pt: 70 °C |
MDL Number:
MFCD00100900 CAS Number: 23761-23-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...