Octenidine dihydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 1,1'-(Decane-1,10-diyl)bis(N-octylpyridin-4(1H)-imine) dihydrochloride

O0388-5G O0388-1G
TCO0388-5GEA 172.22 CAD
TCO0388-5G TCO0388-1G
Octenidine dihydrochloride ≥98.0%
Octenidine dihydrochloride

CAS Number: 70775-75-6
MDL Number: MFCD01938808
Molecular Formula: C36H62N4
Molecular Weight: 623.84
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 214

Formula: C₃₆H₆₂N₄
MW: 623.84 g/mol
Melting Pt: 214 °C
MDL Number: MFCD01938808
CAS Number: 70775-75-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ee3c093dafde7d3157e4f4f96a41811f7b65e37&vid=1ee3c093dafde7d3157e4f4f96a41811f7b65e37&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR