3,4'-Oxydiphthalic anhydride ≥98.0%

Supplier: TCI America

O0384-1G O0384-25G
TCO0384-1GEA 36.34 CAD
TCO0384-1G TCO0384-25G TCO0384-5G
3,4'-Oxydiphthalic anhydride ≥98.0%
3,4'-Oxydiphthalic anhydride

CAS Number: 50662-95-8
Molecular Formula: C16H6O7
Molecular Weight: 310.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 180
Storage Temperature: <0°C

Formula: C₁₆H₆O₇
MW: 310.22 g/mol
Melting Pt: 180 °C
CAS Number: 50662-95-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7263f64defe5f38ce354f531bf889cb038822c10&vid=7263f64defe5f38ce354f531bf889cb038822c10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR