5-n-Octyl-2,2'-bithiophene ≥97.0% (by GC)

Supplier: TCI America

O0383-1G O0383-5G
TCO0383-1GEA 271.13 CAD
TCO0383-1G TCO0383-5G
5-n-Octyl-2,2'-bithiophene ≥97.0% (by GC)
5-n-Octyl-2,2'-bithiophene

CAS Number: 93164-73-9
Molecular Formula: C16H22S2
Molecular Weight: 278.47
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 29
Storage Temperature: <0°C

Formula: C₁₆H₂₂S₂
MW: 278.47 g/mol
Melting Pt: 29 °C
CAS Number: 93164-73-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c7b5ae06daf62ccd622b5552ade3f5ca0566f27&vid=5c7b5ae06daf62ccd622b5552ade3f5ca0566f27&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR