N-n-Octylformamide ≥98.0% (by GC)

Supplier: TCI America

O0379-5G O0379-25G
TCO0379-5GEA 102.7 CAD
TCO0379-5G TCO0379-25G
N-n-Octylformamide ≥98.0% (by GC)
N-n-Octylformamide

CAS Number: 6282-06-0
Molecular Formula: C9H19NO
Molecular Weight: 157.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 291

Formula: C₉H₁₉NO
Boiling Pt: 291 °C
CAS Number: 6282-06-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a110280a0b5b70c8c85c632b6edae494d80db68&vid=2a110280a0b5b70c8c85c632b6edae494d80db68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR