Oxyresveratrol ≥96.0% (by GC)
Supplier: TCI America
Synonyms:
2,3',4,5'-Tetrahydroxy-trans-stilbene, (E)-4-(3,5-Dihydroxystyryl)benzene-1,3-diol
CAS Number: 29700-22-9
MDL Number: MFCD11974969
Molecular Formula: C14H12O4
Molecular Weight: 244.25
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 197
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 2,3',4,5'-Tetrahydroxy-trans-stilbene
Formula:
C₁₄H₁₂O₄ MW: 244.25 g/mol Melting Pt: 197 °C |
MDL Number:
MFCD11974969 CAS Number: 29700-22-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...