Octadecylphosphonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

O0371-1G O0371-5G
TCO0371-1GEA 150.1 CAD
TCO0371-1G TCO0371-5G
Octadecylphosphonic acid ≥98.0% (by titrimetric analysis)
Octadecylphosphonic acid

CAS Number: 4724-47-4
MDL Number: MFCD00015839
Molecular Formula: C18H39O3P
Molecular Weight: 334.48
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 99

Formula: C₁₈H₃₉O₃P
Melting Pt: 99 °C
MDL Number: MFCD00015838
CAS Number: 4724-47-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a711920221a5d07665875bb880547d04096d7c7&vid=0a711920221a5d07665875bb880547d04096d7c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR