(2S,3aS,7aS)-2-Carboxyoctahydroindole ≥98.0%

Supplier: TCI America

Synonyms: (2S,3aS,7aS)-Octahydro-1H-indole-2-carboxylic acid, H-Oic-OH

O0370-1G O0370-5G
TCO0370-1GEA 81.05 CAD
TCO0370-1G TCO0370-5G
(2S,3aS,7aS)-2-Carboxyoctahydroindole ≥98.0%
(2S,3aS,7aS)-2-Carboxyoctahydroindole

CAS Number: 80875-98-5
MDL Number: MFCD07782125
Molecular Formula: C9H15NO2
Molecular Weight: 169.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 270
Specific rotation [a]20/D: -48 deg (C=1, MeOH)

Formula: C9H15NO2
MW: 169.22 g/mol
Melting Pt: 270 °C
MDL Number: MFCD07782125
CAS Number: 80875-98-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d73f77881dcd7aea83097b2523114c8e49b1d239&vid=d73f77881dcd7aea83097b2523114c8e49b1d239&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR