Octakis(dimethylsilyloxy)octasilsesquioxane

Supplier: TCI America

Synonyms: 1,3,5,7,9,11,13,15-Octakis(dimethylsilyloxy)pentacyclo[9.5.1.1(3,9).1(5,15).1(7,13)]octasiloxane

O0351-1G O0351-5G
TCO0351-1GEA 131.14 CAD
TCO0351-1G TCO0351-5G
Octakis(dimethylsilyloxy)octasilsesquioxane
Octakis(dimethylsilyloxy)octasilsesquioxane

CAS Number: 125756-69-6
MDL Number: MFCD03701363
Molecular Formula: C16H56O20Si16
Molecular Weight: 1017.96
Form: Crystal
Color: White
Melting point (°C): 250


Other Product Names / Synonyms: 1,3,5,7,9,11,13,15-Octakis(dimethylsilyloxy)pentacyclo[9.5.1.1(3,9).1(5,15).1(7,13)]octasiloxane

Formula: C₁₆H₅₆O₂₀Si₁₆
MW: 1017.96 g/mol
Melting Pt: 250 °C
MDL Number: MFCD03701363
CAS Number: 125756-69-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67d1ff528d5a28393efb5268bc5fc12f04b1556b&vid=67d1ff528d5a28393efb5268bc5fc12f04b1556b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR