1H,1H,5H-Octafluoropentyl acrylate ≥97.0% (by GC) stabilized

Supplier: TCI America

Synonyms: 2,2,3,3,4,4,5,5-Octafluoropentyl acrylate

O0318-25G O0318-5G
TCO0318-25GEA 229.73 CAD
TCO0318-25G TCO0318-5G
1H,1H,5H-Octafluoropentyl acrylate ≥97.0% (by GC) stabilized
1H,1H,5H-Octafluoropentyl acrylate

(stabilized with MEHQ)
CAS Number: 376-84-1
Molecular Formula: C8H6F8O2
Molecular Weight: 286.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 84
Flash Point (°C): 72
Specific Gravity (20/20): 1.49
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid 1H,1H,5H-Octafluoropentyl Ester

Formula: C₈H₆F₈O₂
MW: 286.12 g/mol
Boiling Pt: 84 °C
Density: 1.49 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00039279
CAS Number: 376-84-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f16173aac70c8cb6d8153ccf30ab7381eccabf9f&vid=f16173aac70c8cb6d8153ccf30ab7381eccabf9f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR