(S)-(-)-1-Octyn-3-ol ≥99.0% (by GC)

Supplier: TCI America

O0235-1G O0235-5G
TCO0235-1GEA 374.46 CAD
TCO0235-1G TCO0235-5G
(S)-(-)-1-Octyn-3-ol ≥99.0% (by GC)
(S)-(-)-1-Octyn-3-ol

[omega Side-Chain Unit for PG Synthesis]
CAS Number: 32556-71-1
MDL Number: MFCD00191475
Molecular Formula: C8H14O
Molecular Weight: 126.20
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 64
Specific Gravity (20/20): 0.86
Specific rotation [a]20/D: -10 deg (neat)

Formula: C₈H₁₄O
Boiling Pt: 130 °C
Density: 0.86 (20 °C)
Flash Pt: 64 °C
MDL Number: MFCD00191475
CAS Number: 32556-71-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86b3df753c4500525ef4edfd24d38e0180ab9948&vid=88b7bed14a418795c87336e36b9a902b0afe3071&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR