Octadecyl vinyl ether ≥85.0% (by GC)

Supplier: TCI America

O0019-25ML O0019-500ML
TCO0019-25MLEA 59.46 CAD
TCO0019-25ML TCO0019-500ML
Octadecyl vinyl ether ≥85.0% (by GC)
Octadecyl vinyl ether

CAS Number: 930-02-9
MDL Number: MFCD00026686
Molecular Formula: C20H40O
Molecular Weight: 296.54
Purity/Analysis Method: >85.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 27


Other Product Names / Synonyms: Stearyl Vinyl Ether

Formula: C₂₀H₄₀O
Flash Pt: 110 °C
MDL Number: MFCD00026686
CAS Number: 930-02-9

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR