4-Nitrobenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

N0989-25G N0989-5G
TCN0989-25GEA 529.3 CAD
TCN0989-25G TCN0989-5G
4-Nitrobenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Nitrobenzamidine hydrochloride

CAS Number: 15723-90-7
MDL Number: MFCD00278833
Molecular Formula: C7H7N3O2
Molecular Weight: 201.61
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 286
Storage Temperature: <0°C

Formula: C₇H₇N₃O₂
Melting Pt: 286 °C
MDL Number: MFCD00278833
CAS Number: 15723-90-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9ee37a28eeb672987159c1929f0e2e69102d756&vid=b9ee37a28eeb672987159c1929f0e2e69102d756&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR