4'-Nitrobiphenyl-4-carboxaldehyde ≥98.0% (by GC)
Supplier: TCI America
N0967-1G
N0967-5G
TCN0967-1GEA
261.17
CAD
TCN0967-1G
TCN0967-5G
4'-Nitrobiphenyl-4-carboxaldehyde ≥98.0% (by GC)
4'-Nitrobiphenyl-4-carboxaldehyde
CAS Number: 98648-23-8
MDL Number: MFCD06200931
Molecular Formula: C13H9NO3
Molecular Weight: 227.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 128
Formula:
C₁₃H₉NO₃ Melting Pt: 128 °C |
MDL Number:
MFCD06200931 CAS Number: 98648-23-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...