S-[4-[(4-Nitrobenzyl)oxy]phenethyl]isothiourea methanesulfonate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4-[(4-Nitrobenzyl)oxy]phenethyl Carbamimidothioate Methanesulfonate, 4-((4-Nitrobenzyl)oxy)phenethyl carbamimidothioate methanesulfonate, KB-R 7943, 4-((4-Nitrobenzyl)oxy)phenethyl carbamimidothioate methanesulphonate, Carbamimidothioic acid 4-[(4-Nitrobenzyl)oxy]phenethyl Ester Methanesulphonate, Carbamimidothioic acid 4-[(4-Nitrobenzyl)oxy]phenethyl Ester Methanesulfonate, 4-[(4-Nitrobenzyl)oxy]phenethyl Carbamimidothioate Methanesulphonate

N0966-10MG N0966-50MG
TCN0966-10MGEA 196.71 CAD
TCN0966-10MG TCN0966-50MG
S-[4-[(4-Nitrobenzyl)oxy]phenethyl]isothiourea methanesulfonate ≥98.0% (by HPLC)
S-[4-[(4-Nitrobenzyl)oxy]phenethyl]isothiourea methanesulfonate

CAS Number: 182004-65-5
MDL Number: MFCD00952138
Molecular Formula: C16H17N3O3S
Molecular Weight: 427.49
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Other Pharmaceutical Ingredients, Na+/Ca2+ Exchanger (NCX) Inhibitors
Other Product Names / Synonyms: 4-[(4-Nitrobenzyl)oxy]phenethyl Carbamimidothioate Methanesulfonate, Carbamimidothioic Acid 4-[(4-Nitrobenzyl)oxy]phenethyl Ester Methanesulfonate, KB-R 7943

Formula: C₁₆H₁₇N₃O₃S
MW: 427.49 g/mol
MDL Number: MFCD00952138
CAS Number: 182004-65-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d4fd21e9a6e86e3e90de691e0fe7a9078c550d8&vid=0d4fd21e9a6e86e3e90de691e0fe7a9078c550d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR