Diamond nanoparticles (particle size : <10nm)
Supplier: TCI America
N0962-5G
N0962-1G
TCN0962-5GEA
507.18
CAD
TCN0962-5G
TCN0962-1G
Diamond nanoparticles (particle size : <10nm)
Diamond
Nanodiamonds (Nanocarbon Materials)
Form: Powder
Color: White
MDL Number:
MFCD00211867 CAS Number: 7782-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...