Nafamostat mesylate ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
6-Amidino-2-naphthyl 4-Guanidinobenzoate Bis(methanesulfonate), 4-Guanidinobenzoic acid 6-Amidino-2-naphthyl Ester Bis(methanesulfonate), 4-Guanidinobenzoic acid 6-Amidino-2-naphthyl Ester Bis(methanesulphonate), 6-Carbamimidoylnaphthalen-2-yl 4-guanidinobenzoate dimethanesulfonate, 6-Carbamimidoylnaphthalen-2-yl 4-guanidinobenzoate dimethanesulphonate, 6-Amidino-2-naphthyl 4-Guanidinobenzoate Bis(methanesulphonate)
CAS Number: 82956-11-4
MDL Number: MFCD00941430
Molecular Formula: C19H17N5O2
Molecular Weight: 539.58
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 258
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antithrombotic Ingredients
Other Product Names / Synonyms: 6-Amidino-2-naphthyl 4-Guanidinobenzoate Bis(methanesulfonate), 4-Guanidinobenzoic Acid 6-Amidino-2-naphthyl Ester Bis(methanesulfonate)
Formula:
C₁₉H₁₇N₅O₂ Melting Pt: 258 °C |
MDL Number:
MFCD00941430 CAS Number: 82956-11-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...