7-Nitro-1,2,3,4-tetrahydroquinoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1,2,3,4-Tetrahydro-7-nitroquinoline

N0955-25G N0955-5G
TCN0955-25GEA 370.19 CAD
TCN0955-25G TCN0955-5G
7-Nitro-1,2,3,4-tetrahydroquinoline ≥98.0% (by GC, titration analysis)
7-Nitro-1,2,3,4-tetrahydroquinoline

CAS Number: 30450-62-5
MDL Number: MFCD00496654
Molecular Formula: C9H10N2O2
Molecular Weight: 178.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 63

Formula: C₉H₁₀N₂O₂
MW: 178.19 g/mol
Melting Pt: 63 °C
MDL Number: MFCD00496654
CAS Number: 30450-62-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e49f1e4da00e18e836fc55cc6d9c3cd160c51d14&vid=e49f1e4da00e18e836fc55cc6d9c3cd160c51d14&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR