4-Nitro-DL-phenylalanine hydrate 98,0

Supplier: TCI America

Synonyms: H-DL-Phe(4-NO2)-OH hydrate, 2-Amino-3-(4-nitrophenyl)propionic acid hydrate

N0952-25G N0952-5G
TCN0952-25GEA 404.95 CAD
TCN0952-25G TCN0952-5G
4-Nitro-DL-phenylalanine hydrate 98,0
4-Nitro-DL-phenylalanine hydrate

CAS Number: 207569-25-3
MDL Number: MFCD00007384
Molecular Formula: C9H10N2O4
Molecular Weight: 210.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 230


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: 2-Amino-3-(4-nitrophenyl)propionic Acid, H-DL-Phe(4-NO2)-OH

Formula: C₉H₁₀N₂O₄
MW: 210.19 g/mol (anhy)
Melting Pt: 230 °C
MDL Number: MFCD00007384
CAS Number: 207569-25-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f864d89d554851adec7edc2c0775d1701055aaac&vid=f864d89d554851adec7edc2c0775d1701055aaac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR