Naphthacene ≥98.0% (by HPLC), purified by sublimation

Supplier: TCI America

N0951-1G N0951-200MG
TCN0951-1GEA 445.56 CAD
TCN0951-1G TCN0951-200MG
Naphthacene ≥98.0% (by HPLC), purified by sublimation
Naphthacene

CAS Number: 92-24-0
MDL Number: MFCD00003702
Molecular Formula: C18H12
Molecular Weight: 228.29
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 357


Product Categories: Materials Chemistry, Organic Transistor Materials, Solar Cell Research Reagents, Nanocarbon Unit Structures (Nanocarbon Materials), Highly Purified Reagents
Other Product Names / Synonyms: 2,3-Benzanthracene, Tetracene

Formula: C₁₈H₁₂
Melting Pt: 357 °C
MDL Number: MFCD00003702
CAS Number: 92-24-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5d82a96436cfea8ab3814f20148479195419b63&vid=b24d1bd6a0283e871df64b9d6f154740ad8e6955&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR