4-(2-Naphthyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
(4-(Naphthalen-2-yl)phenyl)boronic acid
N0946-1G
N0946-200MG
TCN0946-1GEA
132.72
CAD
TCN0946-1G
TCN0946-200MG
4-(2-Naphthyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(2-Naphthyl)phenylboronic acid
CAS Number: 918655-03-5
MDL Number: MFCD09260454
Molecular Formula: C16H13BO2
Molecular Weight: 248.09
Form: Crystal
Color: White
Other Product Names / Synonyms: 4-(2-Naphthyl)benzeneboronic Acid
Formula:
C₁₆H₁₃BO₂ MW: 248.0800 g/mol |
MDL Number:
MFCD09260454 CAS Number: 918655-03-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...