N-(4-Nitrobenzoyl)-β-alanine ≥98.0%
Supplier: TCI America
Synonyms:
3-(4-Nitrobenzamido)propionic Acid
N0942-25G
N0942-5G
TCN0942-25GEA
201.61
CAD
TCN0942-25G
TCN0942-5G
N-(4-Nitrobenzoyl)-β-alanine ≥98.0%
N-(4-Nitrobenzoyl)-beta-alanine
CAS Number: 59642-21-6
MDL Number: MFCD00009794
Molecular Formula: C10H10N2O5
Molecular Weight: 238.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 165
Other Product Names / Synonyms: 3-(4-Nitrobenzamido)propionic Acid
Formula:
C₁₀H₁₀N₂O₅ MW: 238.20 g/mol Melting Pt: 165 °C |
MDL Number:
MFCD00009794 CAS Number: 59642-21-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...