trans-β-Nitrostyrene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (E)-(2-Nitrovinyl)benzene, trans-1-Nitro-2-phenylethylene

N0941-25G N0941-5G
TCN0941-25GEA 344.44 CAD
TCN0941-25G TCN0941-5G
trans-β-Nitrostyrene ≥98.0% (by GC)
trans-β-Nitrostyrene

CAS Number: 5153-67-3
MDL Number: MFCD00007402
Molecular Formula: C8H7NO2
Molecular Weight: 149.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 260
Melting point (°C): 57
Lambda max.: 320 nmStorage Temperature: 0-10°C


Other Product Names / Synonyms: trans-1-Nitro-2-phenylethylene, trans-4-(2-Nitroethenyl)benzene

Formula: C₈H₇NO₂
MW: 149.15 g/mol
Boiling Pt: 260 °C
Melting Pt: 57 °C
MDL Number: MFCD00007402
CAS Number: 5153-67-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=750cd8a4136d0227012e63fb9148e02708056e7c&vid=750cd8a4136d0227012e63fb9148e02708056e7c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR