5-Nitro-2-thiophenecarboxylic acid ≥97.0%

Supplier: TCI America

Synonyms: 5-Nitrothiophene-2-carboxylic acid

N0940-25G N0940-5G
TCN0940-25GEA 404.48 CAD
TCN0940-25G TCN0940-5G
5-Nitro-2-thiophenecarboxylic acid ≥97.0%
5-Nitro-2-thiophenecarboxylic acid

CAS Number: 6317-37-9
MDL Number: MFCD00159552
Molecular Formula: C5H3NO4S
Molecular Weight: 173.14
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 160


Other Product Names / Synonyms: 5-Nitro-2-thenoic Acid

Formula: C₅H₃NO₄S
MW: 173.14 g/mol
Melting Pt: 160 °C
MDL Number: MFCD00159552
CAS Number: 6317-37-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a6eff30da500276942913503d144fe8b65f7fb0&vid=2a6eff30da500276942913503d144fe8b65f7fb0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR