3-Nitro-1H-pyrazole ≥98.0% (by GC)
Supplier: TCI America
N0937-1G
N0937-5G
TCN0937-1GEA
89.27
CAD
TCN0937-1G
TCN0937-5G
3-Nitro-1H-pyrazole ≥98.0% (by GC)
3-Nitro-1H-pyrazole
CAS Number: 26621-44-3
MDL Number: MFCD00159621
Molecular Formula: C3H3N3O2
Molecular Weight: 113.08
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 177
Flash Point (°C): 160
Formula:
C₃H₃N₃O₂ MW: 113.08 Melting Pt: 177 °C Flash Pt: 160 °C |
MDL Number:
MFCD00238787 CAS Number: 26621-44-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...