6-Nitrocoumarin ≥98.0% (by GC)

Supplier: TCI America

N0923-25G N0923-5G
TCN0923-25GEA 248.06 CAD
TCN0923-25G TCN0923-5G
6-Nitrocoumarin ≥98.0% (by GC)
6-Nitrocoumarin

CAS Number: 2725-81-7
MDL Number: MFCD00016973
Molecular Formula: C9H5NO4
Molecular Weight: 191.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 191

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR