Norlaudanosine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 1-(3,4-Dimethoxybenzyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline Hydrochloride, Tetrahydropapaverine Hydrochloride

N0918-25G N0918-5G
TCN0918-25GEA 262.28 CAD
TCN0918-25G TCN0918-5G
Norlaudanosine hydrochloride ≥98.0%
Norlaudanosine hydrochloride

CAS Number: 6429-04-5
MDL Number: MFCD00035267
Molecular Formula: C20H25NO4
Molecular Weight: 379.88
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 218


Product Categories: Bioscience, Isoquinoline Alkaloids
Other Product Names / Synonyms: Tetrahydropapaverine Hydrochloride, 1-(3,4-Dimethoxybenzyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline Hydrochloride

Formula: C₂₀H₂₅NO₄
MW: 379.88 g/mol
Melting Pt: 218 °C
MDL Number: MFCD00035267
CAS Number: 6429-04-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef5debe6725e9303cb2668d7ef6063533e7045c9&vid=ef5debe6725e9303cb2668d7ef6063533e7045c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR